Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcgr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010188.5 | NP_034318.2 | 1 | 84 | ||
GenBank mRNA | AF197930.1 | - | 1 | 79 | ||
AK077227.1 | - | 1 | 44 | |||
AK151557.1 | - | 1 | 44 | |||
AK151795.1 | - | 1 | 44 | |||
AK152875.1 | - | 1 | 44 | |||
AK153384.1 | - | 1 | 44 | |||
AK153449.1 | - | 1 | 44 | |||
AK154892.1 | - | 1 | 45 | |||
AK159919.1 | - | 1 | 33 | |||
AK169887.1 | - | 1 | 44 | |||
AK171289.1 | - | 1 | 45 | |||
AY897419.1 | - | 1 | 33 | |||
AY897420.1 | - | 1 | 29 | |||
AY897421.1 | - | 1 | 33 | |||
AY897422.1 | - | 1 | 33 | |||
AY897423.1 | - | 1 | 33 | |||
AY897424.1 | - | 1 | 28 | |||
AY897425.1 | - | 1 | 38 | |||
AY897426.1 | - | 1 | 38 | |||
AY897427.1 | - | 1 | 38 | |||
AY897428.1 | - | 1 | 42 | |||
AY897429.1 | - | 1 | 33 | |||
BC052819.1 | - | 1 | 38 | 30044058 | ||
BY199827.1 | - | 1 | 84 | |||
M14215.1 | - | 1 | 79 |