Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fhl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289533.1 | NP_001276462.1 | 6 | 875 | ||
NM_010212.4 | NP_034342.1 | 6 | 1024 | |||
GenBank mRNA | AF055889.1 | - | 5 | 752 | ||
AF114381.1 | - | 5 | 752 | |||
AF153340.1 | - | 5 | 796 | |||
AK052936.1 | - | 6 | 876 | |||
AK133708.1 | - | 6 | 996 | |||
AK147024.1 | - | 6 | 1025 | |||
BC021468.1 | - | 6 | 933 | 5066565 | ||
U77040.2 | - | 6 | 914 |