Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flot1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008027.2 | NP_032053.1 | 11 | 1129 | ||
GenBank mRNA | AF145044.1 | - | 10 | 970 | ||
AK150692.1 | - | 11 | 1151 | |||
AK150827.1 | - | 11 | 1078 | |||
AK167323.1 | - | 11 | 1056 | |||
AY167925.1 | - | 11 | 1135 | |||
BC004647.1 | - | 11 | 1139 | 3158071 | ||
U90435.1 | - | 11 | 1129 |