Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276408.1 | NP_001263337.1 | 33 | 5546 | ||
NM_001276409.1 | NP_001263338.1 | 33 | 5546 | |||
NM_001276410.1 | NP_001263339.1 | 32 | 5276 | |||
NM_001276411.1 | NP_001263340.1 | 32 | 5276 | |||
NM_001276412.1 | NP_001263341.1 | 33 | 5546 | |||
NM_001276413.1 | NP_001263342.1 | 32 | 5276 | |||
NM_010233.2 | NP_034363.1 | 34 | 5819 | |||
GenBank mRNA | AK090130.1 | - | 34 | 5770 | ||
AK090135.1 | - | 34 | 5759 | |||
AK133868.1 | - | 8 | 1301 | |||
AK147249.1 | - | 24 | 3896 | |||
AK147315.1 | - | 33 | 5485 | |||
AK147639.1 | - | 33 | 5485 | |||
AK147663.1 | - | 32 | 5214 | |||
AK147683.1 | - | 33 | 5485 | |||
AK147701.1 | - | 32 | 5214 | |||
AK159794.1 | - | 6 | 843 | |||
AK170755.1 | - | 32 | 5244 | |||
AK171339.1 | - | 32 | 5215 | |||
BC025521.1 | - | 4 | 642 | |||
BC036167.1 | - | 7 | 998 | |||
BC085240.1 | - | 5 | 886 | |||
BC138421.1 | - | 32 | 5140 | 8861441 | ||
BC145271.1 | - | 32 | 5140 | 9053825 | ||
X93167.1 | - | 17 | 2874 |