Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276408.1 | NP_001263337.1 | 29, 30 | 4878 | ||
NM_001276409.1 | NP_001263338.1 | 29, 30 | 4878 | |||
NM_001276410.1 | NP_001263339.1 | 29, 30 | 4878 | |||
NM_001276411.1 | NP_001263340.1 | 29, 30 | 4878 | |||
NM_001276412.1 | NP_001263341.1 | 29, 30 | 4878 | |||
NM_001276413.1 | NP_001263342.1 | 29, 30 | 4878 | |||
NM_010233.2 | NP_034363.1 | 30, 31 | 5151 | |||
GenBank mRNA | AK090130.1 | - | 30, 31 | 5102 | ||
AK090135.1 | - | 30, 31 | 5091 | |||
AK133868.1 | - | 4, 5 | 633 | |||
AK147249.1 | - | 20, 21 | 3228 | |||
AK147315.1 | - | 29, 30 | 4817 | |||
AK147639.1 | - | 29, 30 | 4817 | |||
AK147654.1 | - | 29, 30 | 4817 | |||
AK147663.1 | - | 29, 30 | 4816 | |||
AK147683.1 | - | 29, 30 | 4817 | |||
AK147701.1 | - | 29, 30 | 4816 | |||
AK159794.1 | - | 3, 4 | 445 | |||
AK170755.1 | - | 29, 30 | 4846 | |||
AK171339.1 | - | 29, 30 | 4817 | |||
BC036167.1 | - | 4, 5 | 600 | |||
BC085240.1 | - | 2, 3 | 488 | |||
BC138421.1 | - | 29, 30 | 4742 | 8861441 | ||
BC145271.1 | - | 29, 30 | 4742 | 9053825 | ||
X93167.1 | - | 13, 14 | 2206 |