Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fzd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008058.2 | NP_032084.2 | 1 | 505 | ||
GenBank mRNA | BC039165.1 | - | 1 | 192 | ||
BC132543.1 | - | 1 | 505 | 40130820 | ||
BC138062.1 | - | 1 | 505 | 8861081 |
Set Membership: |
G-protein coupled receptor |