Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gata6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010258.3 | NP_034388.2 | 7 | 1926 | ||
GenBank mRNA | AF179425.1 | - | 6 | 1233 | ||
AK142381.1 | - | 7 | 1786 | |||
BG079878.2 | - | 3 | 258 | |||
S82462.1 | - | 6 | 1688 | |||
U51335.1 | - | 5 | 503 |