Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gata6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010258.3 | NP_034388.2 | 4 | 1657 | ||
GenBank mRNA | AF179425.1 | - | 3 | 964 | ||
AK142381.1 | - | 4 | 1517 | |||
S82462.1 | - | 3 | 1419 | |||
U51335.1 | - | 2 | 237 |