Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtpn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008098.4 | NP_032124.1 | 2 | 387 | ||
GenBank mRNA | AK002620.1 | - | 2 | 301 | ||
AK028027.1 | - | 2 | 315 | |||
AK049398.1 | - | 2 | 287 | |||
AK083888.1 | - | 2 | 301 | |||
AK132029.1 | - | 2 | 310 | |||
AK145755.1 | - | 2 | 292 | |||
AK146305.1 | - | 2 | 335 | |||
AK146473.1 | - | 2 | 287 | |||
AK150750.1 | - | 2 | 309 | |||
AK151106.1 | - | 2 | 255 | |||
AK151147.1 | - | 2 | 255 | |||
AK151485.1 | - | 2 | 312 | |||
AK151564.1 | - | 2 | 255 | |||
AK151640.1 | - | 2 | 310 | |||
AK151687.1 | - | 2 | 283 | |||
AK152272.1 | - | 2 | 283 | |||
AK152317.1 | - | 2 | 273 | |||
AK194194.1 | - | 1 | 22 | |||
AK198840.1 | - | 1 | 22 | |||
AK206882.1 | - | 1 | 22 | |||
BC043084.1 | - | 2 | 284 | 6417343 | ||
BC054811.1 | - | 2 | 300 | 6839739 | ||
CA559569.1 | - | 2 | 387 | |||
D78188.1 | - | 2 | 296 | |||
U20290.1 | - | 2 | 318 |