Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gem | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010276.4 | NP_034406.2 | 5 | 764 | ||
GenBank mRNA | AK157655.1 | - | 5 | 766 | ||
AK158393.1 | - | 5 | 766 | |||
AK168401.1 | - | 5 | 765 | |||
AK171336.1 | - | 5 | 702 | |||
BC029668.1 | - | 5 | 740 | 5368352 | ||
U10551.1 | - | 4 | 743 | |||
U13053.1 | - | 5 | 755 |