Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016905.2 | NP_058601.2 | 4 | 593 | ||
GenBank mRNA | AB027012.1 | - | 4 | 549 | ||
AK011103.1 | - | 4 | 594 | |||
AK045694.1 | - | 4 | 567 | |||
AK088151.1 | - | 4 | 594 | |||
AK168716.1 | - | 4 | 553 | |||
AK182284.1 | - | 1 | 57 | |||
BC016602.1 | - | 4 | 593 | 3490224 | ||
BC050151.1 | - | 4 | 564 | 30024030 |