Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013531.4 | NP_038559.2 | 6, 7 | 577 | ||
GenBank mRNA | AF277893.1 | - | 5, 6 | 441 | ||
AF501886.1 | - | 5, 6 | 441 | |||
AK036816.1 | - | 6, 7 | 579 | |||
AK098139.1 | - | 6, 7 | 510 | |||
AK152227.1 | - | 6, 7 | 568 | |||
AK155309.1 | - | 6, 7 | 636 | |||
AK158965.1 | - | 7, 8 | 807 | |||
AK167416.1 | - | 6, 7 | 610 | |||
AK168302.1 | - | 6, 7 | 533 | |||
AK169752.1 | - | 6, 7 | 568 | |||
AK172147.1 | - | 7, 8 | 710 | |||
AK195471.1 | - | 1, 2 | 109 | |||
AK211063.1 | - | 1, 2 | 109 | |||
BC028753.1 | - | 6, 7 | 548 | 1348753 | ||
M63658.1 | - | 5, 6 | 481 | |||
M87286.1 | - | 5, 6 | 481 | |||
S86124.1 | - | 5, 6 | 481 |