Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013531.4 | NP_038559.2 | 3, 4 | 229 | ||
GenBank mRNA | AF277893.1 | - | 2, 3 | 93 | ||
AF501886.1 | - | 2, 3 | 93 | |||
AK036816.1 | - | 3, 4 | 231 | |||
AK098139.1 | - | 3, 4 | 162 | |||
AK152227.1 | - | 3, 4 | 220 | |||
AK155309.1 | - | 3, 4 | 288 | |||
AK158965.1 | - | 4, 5 | 459 | |||
AK165084.1 | - | 3, 4 | 220 | |||
AK167416.1 | - | 3, 4 | 262 | |||
AK168302.1 | - | 3, 4 | 185 | |||
AK169752.1 | - | 3, 4 | 220 | |||
BC028753.1 | - | 3, 4 | 200 | 1348753 | ||
M63658.1 | - | 2, 3 | 133 | |||
M87286.1 | - | 2, 3 | 133 | |||
S86124.1 | - | 2, 3 | 133 |