Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008161.3 | NP_032187.2 | 2 | 225 | ||
GenBank mRNA | AK002219.1 | - | 2 | 154 | ||
AK002262.1 | - | 2 | 154 | |||
AK004942.1 | - | 2 | 154 | |||
AK146760.1 | - | 2 | 154 | |||
BC003339.1 | - | 2 | 137 | 3257333 | ||
BC037027.1 | - | 2 | 172 | 5370640 | ||
BC037134.1 | - | 2 | 151 | |||
BC049235.1 | - | 2 | 141 | 4221025 | ||
BC061950.1 | - | 2 | 129 | 6528998 | ||
BY061328.1 | - | 2 | 226 | |||
BY100863.1 | - | 2 | 227 | |||
BY339672.1 | - | 2 | 227 | |||
CB601893.1 | - | 2 | 157 | |||
U13705.1 | - | 2 | 129 |