Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163465.1 | NP_001156937.1 | 3 | 352 | ||
NM_008164.2 | NP_032190.2 | 3 | 352 | |||
GenBank mRNA | AK143132.1 | - | 3 | 333 | ||
AK220431.1 | - | 3 | 269 | |||
BC004843.1 | - | 3 | 322 | |||
BC052010.1 | - | 3 | 311 | 5717786 | ||
U43194.1 | - | 3 | 246 |