Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gstm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010360.2 | NP_034490.1 | 2 | 165 | ||
GenBank mRNA | AK002750.1 | - | 2 | 97 | ||
AK019317.1 | - | 2 | 96 | |||
AK028098.1 | - | 2 | 181 | |||
BC008206.1 | - | 2 | 119 | 3154170 | ||
U24428.1 | - | 2 | 161 |