Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hlx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008250.2 | NP_032276.1 | 1 | 858 | ||
GenBank mRNA | AK154090.1 | - | 1 | 536 | ||
BB664380.1 | - | 1 | 482 | |||
BC050146.1 | - | 1 | 113 | |||
BC064068.1 | - | 1 | 858 | 1380899 | ||
X58250.1 | - | 1 | 974 |