Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016884.3 | NP_058580.1 | 4 | 518 | ||
GenBank mRNA | AF095257.1 | - | 4 | 472 | ||
AK088245.1 | - | 4 | 495 | |||
AK088678.1 | - | 4 | 490 | |||
AK150399.1 | - | 4 | 492 | |||
AK151446.1 | - | 4 | 495 | |||
AK153053.1 | - | 4 | 495 | |||
DV652725.1 | - | 4 | 542 |