Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016884.3 | NP_058580.1 | 4, 5 | 558 | ||
GenBank mRNA | AF095257.1 | - | 4, 5 | 512 | ||
AK088245.1 | - | 4, 5 | 535 | |||
AK088678.1 | - | 4, 5 | 530 | |||
AK150399.1 | - | 4, 5 | 532 | |||
AK151446.1 | - | 4, 5 | 535 | |||
AK153053.1 | - | 4, 5 | 535 | |||
DV652725.1 | - | 4, 5 | 582 |