Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170981.1 | NP_001164452.1 | 5 | 546 | ||
NM_001170982.1 | NP_001164453.1 | 4 | 520 | |||
NM_001170983.1 | NP_001164454.1 | 5 | 546 | |||
NM_001170984.1 | NP_001164455.1 | 5 | 546 | |||
NM_016884.3 | NP_058580.1 | 5 | 585 | |||
GenBank mRNA | AF095257.1 | - | 5 | 539 | ||
AK010845.1 | - | 5 | 525 | |||
AK011336.1 | - | 5 | 518 | |||
AK012633.2 | - | 5 | 508 | |||
AK019958.1 | - | 5 | 1721 | |||
AK088245.1 | - | 5 | 562 | |||
AK088678.1 | - | 5 | 557 | |||
AK089061.1 | - | 5 | 526 | |||
AK150399.1 | - | 5 | 559 | |||
AK151446.1 | - | 5 | 562 | |||
AK153053.1 | - | 5 | 562 | |||
AK166590.1 | - | 5 | 524 | |||
AK168802.1 | - | 5 | 520 | |||
AK169287.1 | - | 5 | 518 | |||
BC004706.1 | - | 5 | 508 | 3499283 | ||
BC095922.1 | - | 4 | 461 | 4988965 | ||
CK793036.1 | - | 500 | ||||
DV652725.1 | - | 5 | 609 |