Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010461.2 | NP_034591.1 | 1 | 1075 | ||
GenBank mRNA | BC139209.1 | - | 1 | 255 | 8862231 | |
BC139212.1 | - | 2 | 195 | 8862234 | ||
X13721.1 | - | 1 | 1074 |