Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hpn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110252.2 | NP_001103722.1 | 13 | 1475 | ||
NM_001276269.1 | NP_001263198.1 | 12 | 1352 | |||
NM_008281.4 | NP_032307.2 | 12 | 1415 | |||
GenBank mRNA | AF030065.1 | - | 12 | 1374 | ||
AK002694.1 | - | 13 | 1430 | |||
AK156553.1 | - | 12 | 1354 | |||
AY234104.1 | - | 11 | 1480 | |||
BC138809.1 | - | 13 | 1319 | 8861831 | ||
BC145413.1 | - | 13 | 1319 | 9053971 | ||
CB949755.1 | - | 5 | 587 | |||
CJ237993.1 | - | 3 | 292 |