Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hpn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110252.2 | NP_001103722.1 | 11, 12 | 1200 | ||
NM_001276269.1 | NP_001263198.1 | 10, 11 | 1077 | |||
NM_008281.4 | NP_032307.2 | 10, 11 | 1140 | |||
GenBank mRNA | AF030065.1 | - | 10, 11 | 1099 | ||
AK002694.1 | - | 11, 12 | 1155 | |||
AK156553.1 | - | 10, 11 | 1079 | |||
AY234104.1 | - | 9, 10 | 1205 | |||
BC138809.1 | - | 11, 12 | 1044 | 8861831 | ||
BC145413.1 | - | 11, 12 | 1044 | 9053971 | ||
CB949755.1 | - | 3, 4 | 311 |