Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hras | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130443.1 | NP_001123915.1 | 5 | 707 | ||
NM_001130444.1 | NP_001123916.1 | 6 | 789 | |||
NM_008284.2 | NP_032310.2 | 5 | 707 | |||
GenBank mRNA | AJ437023.1 | - | 2 | 133 | ||
AY373386.1 | - | 4 | 501 | |||
BC011083.1 | - | 5 | 635 | 3152667 | ||
BC050837.1 | - | 4 | 1360 | |||
BC061885.1 | - | 663 | ||||
BI731283.1 | - | 355 | ||||
BU924724.1 | - | 3 | 343 |