Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsd17b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008291.3 | NP_032317.2 | 6, 7 | 547 | ||
GenBank mRNA | BB621153.1 | - | 6, 7 | 547 | ||
U66827.1 | - | 6, 7 | 532 |