Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsd3b6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013821.3 | NP_038849.2 | 4 | 722 | ||
GenBank mRNA | AB109387.1 | - | 3 | 398 | ||
AF031170.1 | - | 3 | 462 | |||
AK142267.1 | - | 4 | 724 | |||
AY046512.1 | - | 4 | 713 | |||
BC053501.1 | - | 4 | 708 | 30018625 |