Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002012.1 | NP_001002012.1 | 2 | 1704 | ||
NM_008301.4 | NP_032327.2 | 1 | 1749 | |||
GenBank mRNA | AK132261.1 | - | 1 | 1711 | ||
BC004714.1 | - | 1 | 1716 | 3499661 | ||
BC052350.1 | - | 2 | 1704 | 6772789 |