Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF128210.1 | - | 5 | 468 | ||
AF128211.1 | - | 5 | 468 | |||
AF128212.1 | - | 6 | 395 | |||
AF128213.1 | - | 6 | 395 | |||
AK153729.1 | - | 2 | 525 | |||
BC116851.1 | - | 6 | 399 | |||
BC116855.1 | - | 6 | 399 | |||
M86672.1 | - | 6 | 497 |