Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il1rn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039701.3 | NP_001034790.1 | 3 | 309 | ||
NM_001159562.1 | NP_001153034.1 | 4 | 238 | |||
NM_031167.5 | NP_112444.1 | 4 | 293 | |||
GenBank mRNA | AF001795.1 | - | 4 | 294 | ||
AK076296.1 | - | 4 | 294 | |||
AK089222.1 | - | 3 | 264 | |||
AK154601.1 | - | 3 | 264 | |||
AK155263.1 | - | 3 | 264 | |||
AK155641.1 | - | 3 | 310 | |||
AK155756.1 | - | 3 | 264 | |||
AK155798.1 | - | 3 | 264 | |||
AK155846.1 | - | 3 | 264 | |||
AK155888.1 | - | 3 | 306 | |||
AK155908.1 | - | 3 | 264 | |||
AK156034.1 | - | 3 | 264 | |||
AK157157.1 | - | 3 | 302 | |||
AK170378.1 | - | 4 | 299 | |||
AK171037.1 | - | 4 | 242 | |||
AK171385.1 | - | 3 | 297 | |||
BC042532.1 | - | 4 | 270 | 4486098 | ||
BY206809.1 | - | 4 | 239 | |||
M57525.1 | - | 3 | 242 | |||
M64404.1 | - | 3 | 324 | |||
M74294.1 | - | 3 | 275 | |||
S64082.1 | - | 3 | 161 |