Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inhbb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008381.3 | NP_032407.1 | 2 | 2086 | ||
GenBank mRNA | AK132352.1 | - | 2 | 2088 | ||
BC024067.1 | - | 1 | 238 | |||
BC048845.1 | - | 2 | 792 | |||
X69620.1 | - | 1 | 57 | |||
X83376.1 | - | 1 | 457 |