Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F11r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172647.2 | NP_766235.1 | 3, 4 | 312 | ||
GenBank mRNA | AK033574.1 | - | 3, 4 | 313 | ||
BC021876.1 | - | 3, 4 | 255 | 3985305 | ||
CT010347.1 | - | 3, 4 | 232 |