Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010623.4 | NP_034753.1 | 6, 7 | 1475 | ||
GenBank mRNA | AB008867.1 | - | 6, 7 | 1473 | ||
AK046890.1 | - | 6, 7 | 1228 |