Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008451.2 | NP_032477.2 | 7 | 1156 | ||
GenBank mRNA | AF055666.1 | - | 7 | 989 | ||
AK141439.1 | - | 7 | 1086 | |||
BC014845.1 | - | 7 | 1000 | 2599082 | ||
BM963360.1 | - | 3 | 327 |