Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kras | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK155064.1 | - | 2 | 284 | ||
AY487807.1 | - | 1 | 107 | |||
BC004642.1 | - | 2 | 271 | 3158212 | ||
BC010202.1 | - | 2 | 271 | 3158212 | ||
BC119495.1 | - | 3 | 147 | 40052024 | ||
BC141051.1 | - | 1 | 109 | 9055663 |