Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159374.2 | NP_001152846.2 | 1 | 280 | ||
GenBank mRNA | BC117553.1 | - | 1 | 170 | 40104816 | |
BC118064.1 | - | 1 | 170 | 40104817 | ||
X75649.1 | - | 1 | 170 |