Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ldhc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013580.4 | NP_038608.1 | 4 | 411 | ||
GenBank mRNA | AK007058.1 | - | 4 | 412 | ||
AK161399.1 | - | 4 | 399 | |||
BC049602.1 | - | 4 | 415 | 6774010 | ||
JQ173119.1 | - | 4 | 388 | |||
JQ173120.1 | - | 4 | 388 | |||
L10389.1 | - | 3 | 380 | |||
M17587.1 | - | 3 | 380 | |||
X04752.1 | - | 4 | 419 |