Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lfng | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008494.3 | NP_032520.1 | 7 | 1098 | ||
GenBank mRNA | AF015768.1 | - | 7 | 1127 | ||
AK153283.1 | - | 7 | 1099 | |||
BC115680.1 | - | 6 | 764 | 40104178 | ||
BC138600.1 | - | 7 | 1119 | 8861620 | ||
BC138601.1 | - | 7 | 1119 | 8861621 | ||
U94351.1 | - | 7 | 1063 |