Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mela | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK090120.1 | - | 6767 | |||
AK090126.1 | - | 6770 | ||||
AK090128.1 | - | 6771 | ||||
AK166372.1 | - | 1466 | ||||
BC113756.1 | - | 1199 | 40047695 | |||
D10049.1 | - | 1258 | ||||
U63133.1 | - | 6765 | ||||
X62670.1 | - | 38 |
Gene Symbol: Kcnma1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | GU441834.1 | - | 1507 |