Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010810.4 | NP_034940.2 | 5 | 782 | ||
GenBank mRNA | AK144515.1 | - | 5 | 800 | ||
AY622968.1 | - | 1 | 157 | |||
BC119057.1 | - | 5 | 782 | 8733806 | ||
BC120655.1 | - | 5 | 782 | 40129578 | ||
L36244.1 | - | 5 | 807 |