Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mstn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010834.2 | NP_034964.1 | 3 | 990 | ||
GenBank mRNA | AK132415.1 | - | 3 | 1015 | ||
AK142222.1 | - | 3 | 1014 | |||
BC103676.1 | - | 3 | 990 | 40047210 | ||
BC103677.1 | - | 3 | 990 | 40047209 | ||
BC103678.1 | - | 3 | 990 | 40047208 | ||
BC105674.1 | - | 3 | 990 | 40047211 | ||
U84005.1 | - | 3 | 990 |