Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198635.1 | NP_001185564.1 | 14, 15 | 1910 | ||
NM_008635.2 | NP_032661.2 | 14, 15 | 1886 | |||
GenBank mRNA | AB098611.1 | - | 14, 15 | 1792 | ||
AK014575.1 | - | 14, 15 | 1803 | |||
AK133267.1 | - | 14, 15 | 2335 | |||
BC052637.1 | - | 14, 15 | 1846 | 6467153 | ||
Y15197.1 | - | 14, 15 | 1792 |