Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mup4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008648.1 | NP_032674.1 | 1 | 131 | ||
GenBank mRNA | BC012227.1 | - | 1 | 118 | 4190774 | |
EU882229.1 | - | 1 | 65 | |||
M16358.1 | - | 1 | 131 |