Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mybl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290397.1 | NP_001277326.1 | 6 | 835 | ||
NM_008651.3 | NP_032677.2 | 6 | 835 | |||
GenBank mRNA | BC101945.1 | - | 6 | 579 | ||
BC103533.1 | - | 6 | 580 | 40039678 | ||
BC103534.1 | - | 6 | 579 | |||
L35261.1 | - | 6 | 834 | |||
X82327.1 | - | 6 | 580 |