Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039545.2 | NP_001034634.2 | 28, 29 | 3993 | ||
GenBank mRNA | BC008538.1 | - | 27, 28 | 3932 | 3495412 |