Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ogg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010957.4 | NP_035087.3 | 3 | 754 | ||
GenBank mRNA | AF000669.1 | - | 3 | 632 | ||
AF003596.1 | - | 3 | 719 | |||
AK146870.1 | - | 3 | 754 | |||
AK165875.1 | - | 3 | 706 | |||
AK167456.1 | - | 3 | 814 | |||
AK181469.1 | - | 2 | 368 | |||
BC138733.1 | - | 3 | 710 | 8861755 | ||
BC138736.1 | - | 3 | 710 | 8861758 | ||
U88621.1 | - | 3 | 722 | |||
U96711.1 | - | 3 | 719 | |||
Y11247.1 | - | 3 | 685 | |||
Y13479.1 | - | 3 | 755 |