Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011035.2 | NP_035165.2 | 11, 12 | 1340 | ||
GenBank mRNA | AF082077.1 | - | 11, 12 | 1305 | ||
BC055740.1 | - | 8, 9 | 988 | |||
BC060157.1 | - | 8, 9 | 988 | |||
BC079838.1 | - | 11, 12 | 1325 | |||
BC145698.1 | - | 12, 13 | 1264 |