Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008799.2 | NP_032825.2 | 3 | 656 | ||
GenBank mRNA | AK045985.1 | - | 3 | 657 | ||
AK182333.1 | - | 2 | 139 | |||
DQ906042.1 | - | 636 | ||||
U10903.1 | - | 3 | 636 |
Gene Symbol: Tbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DQ906043.1 | - | 8705 |