Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008799.2 | NP_032825.2 | 5 | 805 | ||
GenBank mRNA | AK045985.1 | - | 5 | 806 | ||
AK138184.1 | - | 1627 | ||||
AK182333.1 | - | 4 | 288 | |||
U10903.1 | - | 5 | 785 |
Gene Symbol: Tbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DQ906043.1 | - | 5421 |