Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011051.3 | NP_035181.1 | 3 | 312 | ||
GenBank mRNA | AK008610.1 | - | 3 | 236 | ||
AK028223.1 | - | 3 | 267 | |||
AK078044.1 | - | 3 | 240 | |||
AK147979.1 | - | 3 | 270 | |||
AK150335.1 | - | 3 | 230 | |||
BC040079.1 | - | 3 | 263 | 3156800 | ||
BU604816.1 | - | 3 | 315 | |||
U49112.1 | - | 3 | 249 | |||
X14938.1 | - | 2 | 79 |