Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011051.3 | NP_035181.1 | 4 | 440 | ||
GenBank mRNA | AK008610.1 | - | 4 | 364 | ||
AK028223.1 | - | 4 | 395 | |||
AK078044.1 | - | 4 | 368 | |||
AK147979.1 | - | 4 | 398 | |||
AK150335.1 | - | 4 | 358 | |||
BC040079.1 | - | 4 | 391 | 3156800 | ||
BU604816.1 | - | 4 | 443 | |||
U49112.1 | - | 4 | 377 | |||
X14938.1 | - | 3 | 207 |